The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145853.1 | NP_001139325.1 | 2 - 3 | 404 | 81 |
NM_006005.3 | NP_005996.2 | 2 - 3 | 408 | 81 | |
XM_017008586.1 | XP_016864075.1 | 3 - 4 | 387 | 81 | |
GenBank mRNA | AF084481.1 | - | 2 - 3 | 408 | 81 |
AK312897.1 | - | 2 - 3 | 337 | 81 | |
BC030130.2 | - | 2 - 3 | 338 | 81 | |
KU178395.1 | - | 1 - 2 | 238 | 81 | |
Y18064.1 | - | 2 - 3 | 396 | 81 |