The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284351.2 | NP_001271280.1 | 4 - 5 | 872 | 87 |
NM_004174.3 | NP_004165.2 | 4 - 5 | 872 | 87 | |
GenBank mRNA | BC101669.1 | - | 4 - 5 | 768 | 87 |
BC101671.1 | - | 4 - 5 | 768 | 87 | |
BC143328.1 | - | 4 - 5 | 768 | 87 | |
U28043.1 | - | 4 - 5 | 770 | 87 |