Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSPC1
Entrez Gene ID
55269
Gene Name
paraspeckle component 1
Gene Aliases
PSP1
Chromosome Location
Chr.13: 19674752 - 19783047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042414.2 NP_001035879.1 4 - 5 912 71
NR_003272.2 3 - 4 1037 71
NR_044998.1 3 - 4 1037 71
XM_006719844.3 XP_006719907.1 3 - 4 1065 71
XM_011535137.1 XP_011533439.1 3 - 4 1024 71
XM_011535138.2 XP_011533440.1 3 - 4 1024 71
XM_011535139.1 XP_011533441.1 3 - 4 1024 71
XM_011535140.1 XP_011533442.1 3 - 4 1024 71
XM_011535141.2 XP_011533443.1 3 - 4 1024 71
XM_011535142.2 XP_011533444.1 3 - 4 1024 71
XM_017020650.1 XP_016876139.1 3 - 4 1024 71
GenBank mRNA AF448795.1 - 3 - 4 775 71
AF449627.1 - 3 - 4 775 71
AK001817.1 - 3 - 4 960 71
AK301554.1 - 5 - 6 737 71
AL834505.1 - 2 - 3 204 71
BC014184.2 - 3 - 4 930 71
BQ685330.1 - 2 - 3 217 71
CR457272.1 - 3 - 4 667 71

View Full Product Details