Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
Chromosome Location
Chr.20: 3667973 - 3682131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282447.2 NP_001269376.1 13 - 14 2090 134
NM_025220.4 NP_079496.1 13 - 14 2090 134
NM_153202.3 NP_694882.1 13 - 14 2090 134
XM_005260843.1 XP_005260900.1 13 - 14 1547 134
XM_006723639.1 XP_006723702.1 13 - 14 1547 134
XM_006723640.1 XP_006723703.1 13 - 14 1538 134
XM_011529366.1 XP_011527668.1 13 - 14 1446 134
XM_011529367.1 XP_011527669.1 13 - 14 1407 134
XM_011529368.1 XP_011527670.1 13 - 14 1547 134
XM_011529370.1 XP_011527672.1 13 - 14 1547 134
XM_011529373.1 XP_011527675.1 12 - 13 1521 134
GenBank mRNA AB055891.1 - 13 - 14 1491 134
AF466287.1 - 13 - 14 1409 134
AK123015.1 - 9 - 10 2114 134
AK126487.1 - 7 - 8 3183 134
AL117415.1 - 3 - 4 1043 134
AY358314.1 - 13 - 14 1517 134
BC125113.1 - 11 - 12 1091 134

View Full Product Details