Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
Chromosome Location
Chr.20: 3667973 - 3682131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153202.3 NP_694882.1 16 - 17 2592 61
XM_011529368.1 XP_011527670.1 16 - 17 2049 61
XM_011529370.1 XP_011527672.1 16 - 17 2017 61
GenBank mRNA AK226148.1 - 7 - 8 3734 61
AL117415.1 - 6 - 7 1545 61

View Full Product Details