Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
Chromosome Location
Chr.20: 3667973 - 3682131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282447.2 NP_001269376.1 18 - 19 2777 63
NM_025220.4 NP_079496.1 18 - 19 2777 63
NM_153202.3 NP_694882.1 17 - 18 2699 63
XM_005260843.1 XP_005260900.1 18 - 19 2234 63
XM_006723639.1 XP_006723702.1 18 - 19 2234 63
XM_006723640.1 XP_006723703.1 18 - 19 2225 63
XM_011529366.1 XP_011527668.1 18 - 19 2133 63
XM_011529367.1 XP_011527669.1 18 - 19 2094 63
XM_011529368.1 XP_011527670.1 17 - 18 2156 63
XM_011529373.1 XP_011527675.1 17 - 18 2208 63
GenBank mRNA AB055891.1 - 18 - 19 2178 63
AF466287.1 - 18 - 19 2096 63
AK092627.1 - 4 - 5 982 63
AK095953.1 - 10 - 11 976 63
AK123015.1 - 14 - 15 2801 63
AK126487.1 - 12 - 13 3778 63
AK126498.1 - 6 - 7 3534 63
AK226148.1 - 8 - 9 3841 63
AL117415.1 - 7 - 8 1652 63
AY358314.1 - 18 - 19 2204 63
BC125112.1 - 6 - 7 1242 63
BC125113.1 - 16 - 17 1778 63

View Full Product Details