The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029939.3 | NP_001025110.2 | 13 - 14 | 3383 | 94 |
NM_001167583.2 | NP_001161055.1 | 12 - 13 | 3224 | 94 | |
NM_001174157.1 | NP_001167628.1 | 11 - 12 | 3115 | 94 | |
NM_001174158.1 | NP_001167629.1 | 12 - 13 | 3224 | 94 | |
NM_001289394.1 | NP_001276323.1 | 13 - 14 | 3460 | 94 | |
NM_020863.3 | NP_065914.2 | 12 - 13 | 3301 | 94 | |
NR_110323.1 | 13 - 14 | 3467 | 94 | ||
XM_011517203.1 | XP_011515505.1 | 12 - 13 | 3198 | 94 | |
XM_011517204.2 | XP_011515506.1 | 11 - 12 | 3021 | 94 | |
XM_011517206.1 | XP_011515508.1 | 12 - 13 | 3222 | 94 | |
XM_017013716.1 | XP_016869205.1 | 11 - 12 | 3096 | 94 | |
GenBank mRNA | AB040918.1 | - | 10 - 11 | 2699 | 94 |
AB167738.1 | - | 12 - 13 | 3290 | 94 | |
AB167739.1 | - | 13 - 14 | 3449 | 94 | |
AB167740.1 | - | 13 - 14 | 3376 | 94 | |
AK300133.1 | - | 13 - 14 | 2910 | 94 | |
AK300324.1 | - | 12 - 13 | 3220 | 94 | |
AK300471.1 | - | 12 - 13 | 3286 | 94 | |
AK301413.1 | - | 2 - 3 | 479 | 94 | |
AL832066.1 | - | 13 - 14 | 3467 | 94 | |
BC012989.2 | - | 12 - 13 | 3215 | 94 | |
BC025423.2 | - | 10 - 11 | 2893 | 94 | |
BC039059.1 | - | 7 - 8 | 985 | 94 | |
BC046180.1 | - | 2 - 3 | 225 | 94 | |
BC098405.1 | - | 12 - 13 | 3224 | 94 | |
BC101766.1 | - | 12 - 13 | 3214 | 94 | |
BC101768.1 | - | 12 - 13 | 3214 | 94 | |
BC143519.1 | - | 11 - 12 | 3029 | 94 |