The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029939.3 | NP_001025110.2 | 7 - 8 | 2516 | 78 |
NM_001167583.2 | NP_001161055.1 | 6 - 7 | 2357 | 78 | |
NM_001174157.1 | NP_001167628.1 | 5 - 6 | 2248 | 78 | |
NM_001174158.1 | NP_001167629.1 | 6 - 7 | 2357 | 78 | |
NM_001289394.1 | NP_001276323.1 | 7 - 8 | 2593 | 78 | |
NM_020863.3 | NP_065914.2 | 6 - 7 | 2434 | 78 | |
NR_110323.1 | 6 - 7 | 2434 | 78 | ||
XM_011517203.1 | XP_011515505.1 | 6 - 7 | 2331 | 78 | |
XM_011517204.2 | XP_011515506.1 | 5 - 6 | 2154 | 78 | |
XM_011517206.1 | XP_011515508.1 | 6 - 7 | 2355 | 78 | |
XM_017013716.1 | XP_016869205.1 | 5 - 6 | 2229 | 78 | |
GenBank mRNA | AB040918.1 | - | 4 - 5 | 1832 | 78 |
AB167738.1 | - | 6 - 7 | 2423 | 78 | |
AB167739.1 | - | 7 - 8 | 2582 | 78 | |
AB167740.1 | - | 7 - 8 | 2509 | 78 | |
AB167741.1 | - | 6 - 7 | 2350 | 78 | |
AK300133.1 | - | 7 - 8 | 2043 | 78 | |
AK300324.1 | - | 6 - 7 | 2353 | 78 | |
AK300471.1 | - | 6 - 7 | 2419 | 78 | |
AL832066.1 | - | 6 - 7 | 2434 | 78 | |
BC012989.2 | - | 6 - 7 | 2348 | 78 | |
BC025423.2 | - | 4 - 5 | 2026 | 78 | |
BC039059.1 | - | 1 - 2 | 118 | 78 | |
BC098405.1 | - | 6 - 7 | 2357 | 78 | |
BC101766.1 | - | 6 - 7 | 2347 | 78 | |
BC101768.1 | - | 6 - 7 | 2347 | 78 | |
BC143519.1 | - | 5 - 6 | 2162 | 78 |