Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
Chromosome Location
Chr.20: 3667973 - 3682131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282447.2 NP_001269376.1 16 - 17 2596 74
NM_025220.4 NP_079496.1 16 - 17 2596 74
XM_005260843.1 XP_005260900.1 16 - 17 2053 74
XM_006723639.1 XP_006723702.1 16 - 17 2053 74
XM_006723640.1 XP_006723703.1 16 - 17 2044 74
XM_011529366.1 XP_011527668.1 16 - 17 1952 74
XM_011529367.1 XP_011527669.1 16 - 17 1913 74
XM_011529373.1 XP_011527675.1 15 - 16 2027 74
GenBank mRNA AB055891.1 - 16 - 17 1997 74
AF466287.1 - 16 - 17 1915 74
AK092627.1 - 2 - 3 801 74
AK095953.1 - 8 - 9 795 74
AK123015.1 - 12 - 13 2620 74
AK126487.1 - 10 - 11 3597 74
AY358314.1 - 16 - 17 2023 74
BC125113.1 - 14 - 15 1597 74

View Full Product Details