The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146256.1 | NP_001139728.1 | 8 - 9 | 1310 | 82 |
NM_144969.2 | NP_659406.1 | 9 - 10 | 1337 | 82 | |
XM_006724624.3 | XP_006724687.1 | 9 - 10 | 874 | 82 | |
XM_017029296.1 | XP_016884785.1 | 9 - 10 | 874 | 82 | |
XM_017029297.1 | XP_016884786.1 | 8 - 9 | 759 | 82 | |
XM_017029298.1 | XP_016884787.1 | 8 - 9 | 747 | 82 | |
GenBank mRNA | AK056374.1 | - | 9 - 10 | 875 | 82 |
AK122885.1 | - | 8 - 9 | 1059 | 82 | |
AK128587.1 | - | 8 - 9 | 845 | 82 | |
AK303896.1 | - | 8 - 9 | 754 | 82 | |
BC103980.1 | - | 9 - 10 | 875 | 82 | |
BC103981.1 | - | 9 - 10 | 875 | 82 | |
BC103982.1 | - | 9 - 10 | 875 | 82 |