Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
Chromosome Location
Chr.10: 87751512 - 87841345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321967.1 NP_001308896.1 9 - 10 1043 134
NM_001321968.1 NP_001308897.1 8 - 9 953 134
NM_001321969.1 NP_001308898.1 10 - 11 1133 134
NM_032810.3 NP_116199.2 9 - 10 1415 134
NR_135914.1 10 - 11 1130 134
XM_005270252.4 XP_005270309.1 9 - 10 1137 134
XM_011540302.1 XP_011538604.1 7 - 8 808 134
XM_017016847.1 XP_016872336.1 9 - 10 1284 134
XM_017016848.1 XP_016872337.1 8 - 9 1287 134
XM_017016849.1 XP_016872338.1 10 - 11 1256 134
XM_017016850.1 XP_016872339.1 10 - 11 1307 134
GenBank mRNA AF361493.1 - 8 - 9 1249 134
AK027506.1 - 6 - 7 643 134
AK075223.1 - 9 - 10 1344 134
AK304297.1 - 6 - 7 716 134
AL834370.1 - 8 - 9 828 134
BC010868.1 - 5 - 6 478 134
BC063530.1 - 8 - 9 874 134
BC073998.1 - 9 - 10 1073 134

View Full Product Details