Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRAMD1A
Entrez Gene ID
57655
Gene Name
GRAM domain containing 1A
Gene Aliases
KIAA1533
Chromosome Location
Chr.19: 34994784 - 35026471 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136199.2 NP_001129671.1 14 - 15 1890 60
NM_001320034.1 NP_001306963.1 15 - 16 1911 60
NM_001320035.1 NP_001306964.1 14 - 15 1798 60
NM_001320036.1 NP_001306965.1 16 - 17 2081 60
NM_020895.4 NP_065946.2 15 - 16 1911 60
XM_011527149.1 XP_011525451.1 15 - 16 1908 60
XM_011527153.1 XP_011525455.1 16 - 17 2084 60
XM_011527154.2 XP_011525456.1 15 - 16 1884 60
XM_011527155.1 XP_011525457.1 16 - 17 2162 60
XM_011527156.1 XP_011525458.1 14 - 15 1798 60
XM_017027034.1 XP_016882523.1 15 - 16 1911 60
XM_017027035.1 XP_016882524.1 15 - 16 2028 60
GenBank mRNA AB040966.1 - 13 - 14 1542 60
AK026064.1 - 6 - 7 728 60
AK074827.1 - 12 - 13 1474 60
AK074864.1 - 14 - 15 1800 60
AK074914.1 - 14 - 15 1784 60
AK097215.1 - 16 - 17 2081 60
BC014077.2 - 15 - 16 1899 60
BC041618.1 - 5 - 6 1538 60

View Full Product Details