Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRAMD1A
Entrez Gene ID
57655
Gene Name
GRAM domain containing 1A
Gene Aliases
KIAA1533
Chromosome Location
Chr.19: 34994784 - 35026471 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320034.1 NP_001306963.1 2 - 3 377 63
NM_001320035.1 NP_001306964.1 2 - 3 377 63
NM_001320036.1 NP_001306965.1 3 - 4 550 63
NM_020895.4 NP_065946.2 2 - 3 377 63
XM_011527149.1 XP_011525451.1 2 - 3 377 63
XM_011527153.1 XP_011525455.1 3 - 4 550 63
XM_011527154.2 XP_011525456.1 2 - 3 350 63
XM_011527155.1 XP_011525457.1 3 - 4 628 63
XM_011527156.1 XP_011525458.1 2 - 3 377 63
XM_017027034.1 XP_016882523.1 2 - 3 377 63
XM_017027035.1 XP_016882524.1 2 - 3 494 63
GenBank mRNA AK074864.1 - 2 - 3 379 63
AK097215.1 - 3 - 4 550 63
BC014077.2 - 2 - 3 365 63
DA390646.1 - 2 - 3 384 63

View Full Product Details