The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003884.4 | NP_003875.3 | 13 - 14 | 2449 | 89 |
XM_005265528.4 | XP_005265585.1 | 13 - 14 | 2074 | 89 | |
XM_017007423.1 | XP_016862912.1 | 14 - 15 | 1904 | 89 | |
XM_017007424.1 | XP_016862913.1 | 13 - 14 | 2000 | 89 | |
GenBank mRNA | AL832173.1 | - | 3 - 4 | 362 | 89 |
BC060823.1 | - | 13 - 14 | 2054 | 89 | |
BC070075.1 | - | 13 - 14 | 2119 | 89 | |
U57317.2 | - | 13 - 14 | 2461 | 89 |