The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015141.3 | NP_055956.1 | 2 - 3 | 428 | 112 |
GenBank mRNA | AK292808.1 | - | 2 - 3 | 261 | 112 |
BC028726.2 | - | 2 - 3 | 287 | 112 | |
D42047.1 | - | 2 - 3 | 408 | 112 | |
DA318838.1 | - | 2 - 3 | 428 | 112 |