Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYNC1I2
Entrez Gene ID
1781
Gene Name
dynein cytoplasmic 1 intermediate chain 2
Gene Aliases
DIC74, DNCI2, IC2
Chromosome Location
Chr.2: 171687409 - 171750158 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271785.1 NP_001258714.1 10 - 11 1078 102
NM_001271786.1 NP_001258715.1 8 - 9 1071 102
NM_001271787.1 NP_001258716.1 8 - 9 1054 102
NM_001271788.1 NP_001258717.1 8 - 9 1017 102
NM_001271789.1 NP_001258718.1 8 - 9 1000 102
NM_001271790.1 NP_001258719.1 8 - 9 1017 102
NM_001320882.1 NP_001307811.1 9 - 10 1077 102
NM_001320883.1 NP_001307812.1 9 - 10 1060 102
NM_001320884.1 NP_001307813.1 8 - 9 1000 102
NM_001378.2 NP_001369.1 10 - 11 1095 102
XM_006712347.3 XP_006712410.1 9 - 10 1001 102
XM_017003525.1 XP_016859014.1 9 - 10 1036 102
GenBank mRNA AB209014.1 - 6 - 7 1020 102
AF134477.1 - 8 - 9 889 102
AF250307.1 - 8 - 9 909 102
AK026574.1 - 1 - 1 103 102
AK055491.1 - 10 - 11 1032 102
AK096579.1 - 10 - 11 996 102
AK225759.1 - 8 - 9 963 102
AK298561.1 - 8 - 9 914 102
AK308008.1 - 8 - 9 872 102
AK316119.1 - 8 - 9 934 102
AL137519.1 - 2 - 3 240 102
AL832832.1 - 4 - 5 387 102
AY037160.1 - 8 - 9 925 102
BC015038.2 - 8 - 9 861 102
BC091498.1 - 8 - 9 945 102
BC109375.1 - 8 - 9 902 102
BT007130.1 - 7 - 8 789 102
BX537412.1 - 8 - 9 957 102
DQ786324.1 - 4 - 5 415 102

View Full Product Details