The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127649.2 | NP_001121121.1 | 3 - 4 | 1053 | 60 |
NM_017929.5 | NP_060399.1 | 4 - 5 | 879 | 60 | |
GenBank mRNA | AB089678.1 | - | 3 - 4 | 769 | 60 |
AB103104.1 | - | 3 - 4 | 671 | 60 | |
AB103106.1 | - | 3 - 4 | 670 | 60 | |
AB103107.1 | - | 3 - 4 | 670 | 60 | |
AB103108.1 | - | 3 - 4 | 670 | 60 | |
AB103109.1 | - | 3 - 4 | 670 | 60 | |
AB103110.1 | - | 3 - 4 | 671 | 60 | |
AB242827.1 | - | 3 - 4 | 670 | 60 | |
AB242828.1 | - | 3 - 4 | 670 | 60 | |
AB242829.1 | - | 3 - 4 | 670 | 60 | |
AB242830.1 | - | 3 - 4 | 670 | 60 | |
AK000065.1 | - | 4 - 5 | 879 | 60 | |
AK000702.1 | - | 4 - 5 | 876 | 60 | |
BC016280.1 | - | 4 - 5 | 797 | 60 | |
BC047320.1 | - | 3 - 4 | 1039 | 60 | |
CR456362.1 | - | 3 - 4 | 688 | 60 | |
KU178709.1 | - | 3 - 4 | 670 | 60 |