Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANTXR2
Entrez Gene ID
118429
Gene Name
anthrax toxin receptor 2
Gene Aliases
CMG-2, CMG2, HFS, ISH, JHF
Chromosome Location
Chr.4: 79901617 - 80073472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK055636.1 - 17 - 18 1423 67

View Full Product Details