The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024939.3 | NP_001020110.1 | 1 - 2 | 183 | 64 |
GenBank mRNA | AK304409.1 | - | 1 - 2 | 161 | 64 |
AK308589.1 | - | 1 - 2 | 161 | 64 | |
AK316049.1 | - | 1 - 2 | 132 | 64 | |
CR456373.1 | - | 1 - 2 | 72 | 64 | |
DC303452.1 | - | 1 - 2 | 184 | 64 |