The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001279357.1 | NP_001266286.1 | 5 - 6 | 502 | 140 |
NM_001279358.1 | NP_001266287.1 | 6 - 7 | 550 | 140 | |
NM_001279359.1 | NP_001266288.1 | 6 - 7 | 510 | 140 | |
NM_001279360.1 | NP_001266289.1 | 6 - 7 | 492 | 140 | |
NM_006330.3 | NP_006321.1 | 6 - 7 | 550 | 140 | |
XM_005251127.4 | XP_005251184.1 | 6 - 7 | 350 | 140 | |
XM_017012956.1 | XP_016868445.1 | 5 - 6 | 461 | 140 | |
XM_017012957.1 | XP_016868446.1 | 7 - 8 | 500 | 140 | |
XM_017012958.1 | XP_016868447.1 | 6 - 7 | 427 | 140 | |
XM_017012959.1 | XP_016868448.1 | 7 - 8 | 549 | 140 | |
XM_017012960.1 | XP_016868449.1 | 5 - 6 | 367 | 140 | |
GenBank mRNA | AA307642.1 | - | 6 - 7 | 308 | 140 |
AF052112.1 | - | 6 - 7 | 550 | 140 | |
AF077198.1 | - | 6 - 7 | 361 | 140 | |
AF077199.1 | - | 5 - 6 | 313 | 140 | |
AF081281.1 | - | 6 - 7 | 391 | 140 | |
AF291053.1 | - | 6 - 7 | 374 | 140 | |
AK289400.1 | - | 6 - 7 | 434 | 140 | |
AK313739.1 | - | 6 - 7 | 387 | 140 | |
BC008652.2 | - | 6 - 7 | 438 | 140 | |
BC010397.1 | - | 6 - 7 | 362 | 140 | |
CR457103.1 | - | 6 - 7 | 356 | 140 | |
DA817192.1 | - | 6 - 7 | 334 | 140 | |
DA881616.1 | - | 6 - 7 | 382 | 140 |