Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSR1
Entrez Gene ID
55720
Gene Name
TSR1, ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.17: 2322678 - 2337384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018128.4 NP_060598.3 11 - 12 2861 75
GenBank mRNA AB037822.1 - 11 - 12 2984 75
AK001396.1 - 9 - 10 1503 75
AK026565.1 - 11 - 12 1934 75
BC019090.2 - 10 - 11 1769 75
BC110851.1 - 11 - 12 2439 75
BC126110.1 - 11 - 12 2409 75

View Full Product Details