Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSR1
Entrez Gene ID
55720
Gene Name
TSR1, ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.17: 2322678 - 2337384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018128.4 NP_060598.3 12 - 13 2980 71
GenBank mRNA AB037822.1 - 12 - 13 3103 71
AK001396.1 - 10 - 11 1622 71
AK026565.1 - 12 - 13 2053 71
BC019090.2 - 11 - 12 1888 71
BC110851.1 - 12 - 13 2558 71
BC126110.1 - 12 - 13 2528 71

View Full Product Details