Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC15
Entrez Gene ID
158866
Gene Name
zinc finger DHHC-type containing 15
Gene Aliases
MRX91
Chromosome Location
Chr.X: 75368427 - 75523626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144969.2 NP_659406.1 1 - 2 614 101
XM_006724624.3 XP_006724687.1 1 - 2 151 101
XM_017029296.1 XP_016884785.1 1 - 2 151 101
XM_017029297.1 XP_016884786.1 1 - 2 157 101
XM_017029298.1 XP_016884787.1 1 - 2 145 101
GenBank mRNA AK056374.1 - 1 - 2 152 101
BC103980.1 - 1 - 2 152 101
BC103981.1 - 1 - 2 152 101
BC103982.1 - 1 - 2 152 101

View Full Product Details