Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR81
Entrez Gene ID
124997
Gene Name
WD repeat domain 81
Gene Aliases
CAMRQ2, PPP1R166, SORF-2
Chromosome Location
Chr.17: 1716523 - 1738599 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163673.1 NP_001157145.1 8 - 9 1768 87
NM_001163809.1 NP_001157281.1 8 - 9 5453 87
NM_001163811.1 NP_001157283.1 8 - 9 1904 87
NM_152348.3 NP_689561.2 9 - 10 2405 87
XM_011523651.2 XP_011521953.1 9 - 10 2390 87
GenBank mRNA AK074356.1 - 3 - 4 995 87
AK122972.1 - 8 - 9 2715 87
AK123896.1 - 8 - 9 1895 87
AK127946.1 - 8 - 9 1904 87
AK298567.1 - 8 - 9 1768 87
AK300986.1 - 6 - 7 1385 87
AL834379.1 - 8 - 9 2229 87
BC092513.1 - 8 - 9 2180 87
BC114519.1 - 7 - 8 1663 87
BC114568.1 - 8 - 9 1730 87

View Full Product Details