Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC2A11
Entrez Gene ID
66035
Gene Name
solute carrier family 2 member 11
Gene Aliases
GLUT10, GLUT11
Chromosome Location
Chr.22: 23856703 - 23886112 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024938.3 NP_001020109.1 10 - 11 1432 83
NM_001024939.3 NP_001020110.1 10 - 11 1320 83
NM_001282864.1 NP_001269793.1 9 - 10 1648 83
NM_030807.4 NP_110434.3 11 - 12 1543 83
NR_104247.1 9 - 10 1279 83
GenBank mRNA AB209301.1 - 6 - 7 1905 83
AF443201.1 - 11 - 12 1374 83
AJ271290.1 - 10 - 11 1263 83
AK075467.1 - 10 - 11 1208 83
AK300196.1 - 9 - 10 1648 83
AK314502.1 - 11 - 12 1314 83
BC100808.2 - 10 - 11 1178 83
BC100809.1 - 10 - 11 1178 83
BC100810.1 - 9 - 10 1025 83
CR456373.1 - 10 - 11 1209 83

View Full Product Details