Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSR1
Entrez Gene ID
55720
Gene Name
TSR1, ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.17: 2322678 - 2337384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018128.4 NP_060598.3 1 - 2 1058 108
GenBank mRNA AB037822.1 - 1 - 2 1181 108
AK026565.1 - 1 - 2 131 108
BC110851.1 - 1 - 2 636 108
BC126110.1 - 1 - 2 606 108

View Full Product Details