The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318328.1 | NP_001305257.1 | 7 - 8 | 2453 | 77 |
NM_015045.3 | NP_055860.1 | 7 - 8 | 2471 | 77 | |
XM_011539548.1 | XP_011537850.1 | 7 - 8 | 2521 | 77 | |
XM_017015979.1 | XP_016871468.1 | 6 - 7 | 2368 | 77 | |
XM_017015980.1 | XP_016871469.1 | 6 - 7 | 2311 | 77 | |
XM_017015981.1 | XP_016871470.1 | 4 - 5 | 2058 | 77 | |
GenBank mRNA | AB065003.1 | - | 7 - 8 | 2474 | 77 |
AF479418.1 | - | 8 - 9 | 2149 | 77 | |
AK290138.1 | - | 7 - 8 | 2432 | 77 | |
BC140858.1 | - | 7 - 8 | 2177 | 77 | |
BC150269.1 | - | 8 - 9 | 2322 | 77 | |
D87450.1 | - | 8 - 9 | 2322 | 77 |