The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252078.1 | NP_001239007.1 | 2 - 3 | 290 | 136 |
NM_001252079.1 | NP_001239008.1 | 2 - 3 | 290 | 136 | |
NM_006313.2 | NP_006304.1 | 2 - 3 | 290 | 136 | |
XM_006719718.3 | XP_006719781.1 | 2 - 3 | 288 | 136 | |
XM_017020285.1 | XP_016875774.1 | 2 - 3 | 241 | 136 | |
GenBank mRNA | AB011101.2 | - | 2 - 3 | 215 | 136 |
AF013990.1 | - | 2 - 3 | 161 | 136 | |
AF106069.1 | - | 2 - 3 | 224 | 136 | |
AF153604.1 | - | 2 - 3 | 272 | 136 | |
AK023703.1 | - | 2 - 3 | 233 | 136 | |
AK292337.1 | - | 2 - 3 | 250 | 136 | |
AK295947.1 | - | 2 - 3 | 222 | 136 | |
AK297759.1 | - | 2 - 3 | 226 | 136 | |
BC020688.1 | - | 2 - 3 | 226 | 136 | |
BC063454.1 | - | 2 - 3 | 230 | 136 | |
BC110908.1 | - | 2 - 3 | 223 | 136 | |
BC125123.1 | - | 2 - 3 | 224 | 136 | |
DB451322.1 | - | 2 - 3 | 292 | 136 |