The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006113.4 | NP_006104.4 | 11 - 12 | 1136 | 137 |
XM_005270360.2 | XP_005270417.1 | 10 - 11 | 948 | 137 | |
XM_005270361.1 | XP_005270418.1 | 11 - 12 | 1129 | 137 | |
XM_017000053.1 | XP_016855542.1 | 11 - 12 | 1129 | 137 | |
XM_017000054.1 | XP_016855543.1 | 11 - 12 | 1129 | 137 | |
XM_017000055.1 | XP_016855544.1 | 10 - 11 | 806 | 137 | |
GenBank mRNA | AF067817.1 | - | 11 - 12 | 1119 | 137 |
AF118886.1 | - | 10 - 11 | 940 | 137 | |
AF118887.1 | - | 11 - 12 | 1128 | 137 | |
AK097750.1 | - | 12 - 13 | 1157 | 137 | |
AK295166.1 | - | 11 - 12 | 1066 | 137 | |
BC143969.1 | - | 11 - 12 | 1219 | 137 |