The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185054.1 | NP_001171983.1 | 7 - 8 | 970 | 62 |
NM_001185055.1 | NP_001171984.1 | 6 - 7 | 1097 | 62 | |
NM_001617.3 | NP_001608.1 | 7 - 8 | 1216 | 62 | |
NM_017482.3 | NP_059516.2 | 7 - 8 | 1216 | 62 | |
NM_017488.3 | NP_059522.1 | 7 - 8 | 1216 | 62 | |
XM_011532502.2 | XP_011530804.1 | 7 - 8 | 957 | 62 | |
GenBank mRNA | AB209227.1 | - | 7 - 8 | 1185 | 62 |
AB593080.1 | - | 6 - 7 | 1097 | 62 | |
AF486420.1 | - | 6 - 7 | 773 | 62 | |
AK291007.1 | - | 7 - 8 | 1171 | 62 | |
AK297250.1 | - | 6 - 7 | 1052 | 62 | |
AK309773.1 | - | 6 - 7 | 1051 | 62 | |
BC010237.1 | - | 7 - 8 | 1150 | 62 | |
BC041666.1 | - | 7 - 8 | 1150 | 62 | |
BC051882.1 | - | 7 - 8 | 1143 | 62 | |
BC056881.1 | - | 7 - 8 | 999 | 62 | |
BC065525.1 | - | 7 - 8 | 970 | 62 |