Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA12
Entrez Gene ID
26154
Gene Name
ATP binding cassette subfamily A member 12
Gene Aliases
ARCI4A, ARCI4B, ICR2B, LI2
Chromosome Location
Chr.2: 214931542 - 215138591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015657.3 NP_056472.2 40 - 41 6445 75
NM_173076.2 NP_775099.2 48 - 49 7460 75
NR_103740.1 50 - 51 7540 75
XM_011510951.2 XP_011509253.1 48 - 49 7632 75
GenBank mRNA AF418105.1 - 43 - 44 6582 75
AK096597.1 - 4 - 5 585 75
AK123578.1 - 15 - 16 2000 75
AL080207.1 - 1 - 2 44 75
AY033486.1 - 40 - 41 6445 75
AY219711.1 - 48 - 49 7460 75

View Full Product Details