Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRK4
Entrez Gene ID
2868
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
GPRK2L, GPRK4, GRK4a, IT11
Chromosome Location
Chr.4: 2963504 - 3040760 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004057.1 NP_001004057.1 1 - 2 503 101
NM_182982.2 NP_892027.2 1 - 2 503 101
XM_011513447.2 XP_011511749.1 1 - 2 615 101
XM_011513448.2 XP_011511750.1 1 - 2 615 101
XM_011513450.2 XP_011511752.1 1 - 2 615 101
XM_011513451.2 XP_011511753.1 1 - 2 615 101
XM_011513452.2 XP_011511754.1 1 - 2 615 101
XM_011513453.2 XP_011511755.1 1 - 2 615 101
XM_011513454.2 XP_011511756.1 1 - 2 615 101
XM_011513455.2 XP_011511757.1 1 - 2 615 101
XM_011513456.2 XP_011511758.1 1 - 2 615 101
XM_011513457.2 XP_011511759.1 1 - 2 615 101
XM_017008052.1 XP_016863541.1 1 - 2 615 101
XM_017008053.1 XP_016863542.1 1 - 2 615 101
XM_017008054.1 XP_016863543.1 1 - 2 615 101
XM_017008056.1 XP_016863545.1 1 - 2 615 101
XM_017008062.1 XP_016863551.1 1 - 2 530 101
XM_017008065.1 XP_016863554.1 1 - 2 615 101
GenBank mRNA AK292390.1 - 1 - 2 548 101
BC117320.1 - 1 - 2 391 101
BI465220.1 - 1 - 2 547 101
U33054.1 - 1 - 2 302 101
U33056.1 - 1 - 2 302 101
X75897.1 - 1 - 2 42 101
X97881.1 - 1 - 2 146 101
X98118.1 - 1 - 2 146 101

View Full Product Details