Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNH2
Entrez Gene ID
3757
Gene Name
potassium voltage-gated channel subfamily H member 2
Gene Aliases
ERG-1, ERG1, H-ERG, HERG, HERG1, Kv11.1, LQT2, SQT1
Chromosome Location
Chr.7: 150944956 - 150978314 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000238.3 NP_000229.1 12 - 13 3368 67
NM_172057.2 NP_742054.1 8 - 9 2273 67
XM_011516185.2 XP_011514487.1 11 - 12 2779 67
XM_011516186.2 XP_011514488.1 12 - 13 3173 67
XM_017012195.1 XP_016867684.1 11 - 12 3449 67
XM_017012196.1 XP_016867685.1 12 - 13 2909 67
GenBank mRNA AF363636.1 - 12 - 13 2967 67
AJ512214.1 - 8 - 9 1964 67
BC004311.2 - 4 - 5 822 67
DQ525913.1 - 12 - 13 3368 67
FJ938021.1 - 10 - 11 3800 67
U04270.1 - 12 - 13 3150 67

View Full Product Details