Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C2
Entrez Gene ID
717
Gene Name
complement component 2
Gene Aliases
ARMD14, CO2
Chromosome Location
Chr.6: 31897785 - 31945674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000063.5 NP_000054.2 11 - 12 1734 87
NM_001145903.2 NP_001139375.1 9 - 10 1338 87
NM_001178063.2 NP_001171534.1 7 - 8 884 87
NM_001282457.1 NP_001269386.1 7 - 8 941 87
NM_001282458.1 NP_001269387.1 11 - 12 1762 87
GenBank mRNA AK096258.1 - 8 - 9 1040 87
AK222537.1 - 11 - 12 1486 87
AK222783.1 - 11 - 12 1458 87
AK298311.1 - 7 - 8 884 87
AK298808.1 - 7 - 8 941 87
AK300930.1 - 11 - 12 1500 87
AK307930.1 - 9 - 10 1092 87
AK312581.1 - 11 - 12 1568 87
BC043484.1 - 11 - 12 1469 87
BX537504.1 - 5 - 6 1471 87
X04481.1 - 11 - 12 1494 87

View Full Product Details