Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBM19
Entrez Gene ID
9904
Gene Name
RNA binding motif protein 19
Gene Aliases
-
Chromosome Location
Chr.12: 113816738 - 113966371 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146698.1 NP_001140170.1 4 - 5 524 116
NM_001146699.1 NP_001140171.1 4 - 5 524 116
NM_016196.3 NP_057280.2 4 - 5 524 116
XM_017020280.1 XP_016875769.1 4 - 5 524 116
XM_017020281.1 XP_016875770.1 4 - 5 524 116
GenBank mRNA AB014582.1 - 4 - 5 459 116
AK291444.1 - 4 - 5 475 116
AK314606.1 - 4 - 5 524 116
AL117547.1 - 4 - 5 465 116
BC004289.1 - 4 - 5 463 116
BC006137.1 - 4 - 5 463 116

View Full Product Details