The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146698.1 | NP_001140170.1 | 6 - 7 | 987 | 76 |
NM_001146699.1 | NP_001140171.1 | 6 - 7 | 987 | 76 | |
NM_016196.3 | NP_057280.2 | 6 - 7 | 987 | 76 | |
XM_017020280.1 | XP_016875769.1 | 6 - 7 | 987 | 76 | |
XM_017020281.1 | XP_016875770.1 | 6 - 7 | 987 | 76 | |
GenBank mRNA | AB014582.1 | - | 6 - 7 | 922 | 76 |
AK291444.1 | - | 6 - 7 | 938 | 76 | |
AK314606.1 | - | 6 - 7 | 987 | 76 | |
AL117547.1 | - | 6 - 7 | 928 | 76 | |
BC004289.1 | - | 6 - 7 | 926 | 76 | |
BC006137.1 | - | 6 - 7 | 926 | 76 |