Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP7A
Entrez Gene ID
538
Gene Name
ATPase copper transporting alpha
Gene Aliases
DSMAX, MK, MNK, SMAX3
Chromosome Location
Chr.X: 77910656 - 78050395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000052.6 NP_000043.4 14 - 15 3114 104
NM_001282224.1 NP_001269153.1 13 - 14 2880 104
GenBank mRNA AB117973.1 - 14 - 15 3041 104
L06133.1 - 14 - 15 3058 104

View Full Product Details