The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001870.3 | NP_001861.2 | 6 - 7 | 632 | 149 |
GenBank mRNA | AK313628.1 | - | 6 - 7 | 592 | 149 |
BC012613.1 | - | 6 - 7 | 595 | 149 | |
M27717.1 | - | 6 - 7 | 580 | 149 | |
S40234.1 | - | 3 - 4 | 253 | 149 |