The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019119.4 | NP_061992.3 | 1 - 1 | 3657 | 89 |
GenBank mRNA | AB208959.1 | - | 1 - 1 | 3223 | 89 |
AF217749.1 | - | 1 - 1 | 3475 | 89 |