The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304803.1 | NP_001291732.1 | 4 - 5 | 424 | 58 |
NM_021947.2 | NP_068766.1 | 5 - 6 | 596 | 58 | |
XM_006721565.3 | XP_006721628.1 | 5 - 6 | 628 | 58 | |
XM_006721566.3 | XP_006721629.1 | 5 - 6 | 1806 | 58 | |
XM_011523974.2 | XP_011522276.1 | 5 - 6 | 735 | 58 | |
GenBank mRNA | AF169974.1 | - | 4 - 5 | 533 | 58 |
AK023169.1 | - | 5 - 6 | 585 | 58 | |
AK223120.1 | - | 5 - 6 | 589 | 58 | |
AL834378.1 | - | 3 - 4 | 361 | 58 | |
AY034081.1 | - | 4 - 5 | 517 | 58 | |
BC074728.2 | - | 4 - 5 | 535 | 58 | |
CR457300.1 | - | 4 - 5 | 517 | 58 | |
DA297969.1 | - | 4 - 5 | 413 | 58 |