The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160213.1 | NP_001153685.1 | 12 - 13 | 1986 | 77 |
NM_031293.2 | NP_112583.2 | 11 - 12 | 1808 | 77 | |
XM_011523357.2 | XP_011521659.1 | 11 - 12 | 1913 | 77 | |
XM_011523358.2 | XP_011521660.1 | 11 - 12 | 1853 | 77 | |
XM_011523360.2 | XP_011521662.1 | 11 - 12 | 1876 | 77 | |
XM_011523361.2 | XP_011521663.1 | 11 - 12 | 2351 | 77 | |
XM_011523362.2 | XP_011521664.1 | 10 - 11 | 1658 | 77 | |
GenBank mRNA | AK097465.1 | - | 7 - 8 | 1045 | 77 |
AK122925.1 | - | 12 - 13 | 1986 | 77 | |
AK315004.1 | - | 11 - 12 | 1789 | 77 | |
AL136795.1 | - | 11 - 12 | 1768 | 77 | |
BC028423.2 | - | 11 - 12 | 1810 | 77 |