Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMFBP1
Entrez Gene ID
83449
Gene Name
polyamine modulated factor 1 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 72116696 - 72173908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031293.2 NP_112583.2 3 - 4 338 61
XM_011523357.2 XP_011521659.1 3 - 4 428 61
XM_011523358.2 XP_011521660.1 3 - 4 368 61
XM_011523360.2 XP_011521662.1 3 - 4 391 61
XM_011523361.2 XP_011521663.1 3 - 4 866 61
XM_011523362.2 XP_011521664.1 2 - 3 173 61
GenBank mRNA AK093285.1 - 3 - 4 334 61
AK315004.1 - 3 - 4 319 61
AL136795.1 - 3 - 4 298 61
BC028423.2 - 3 - 4 325 61
DB019901.1 - 3 - 4 428 61
DB082449.1 - 3 - 4 370 61

View Full Product Details