Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMFBP1
Entrez Gene ID
83449
Gene Name
polyamine modulated factor 1 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 72116696 - 72173908 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160213.1 NP_001153685.1 8 - 9 1270 76
NM_031293.2 NP_112583.2 7 - 8 1092 76
XM_011523357.2 XP_011521659.1 7 - 8 1182 76
XM_011523358.2 XP_011521660.1 7 - 8 1122 76
XM_011523360.2 XP_011521662.1 7 - 8 1145 76
XM_011523361.2 XP_011521663.1 7 - 8 1620 76
XM_011523362.2 XP_011521664.1 6 - 7 927 76
GenBank mRNA AK093285.1 - 7 - 8 1088 76
AK097465.1 - 3 - 4 329 76
AK122925.1 - 8 - 9 1270 76
AK315004.1 - 7 - 8 1073 76
AL136795.1 - 7 - 8 1052 76
BC028423.2 - 7 - 8 1079 76

View Full Product Details