The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006741.3 | NP_006732.3 | 6 - 7 | 675 | 64 |
XM_005268995.4 | XP_005269052.1 | 5 - 6 | 499 | 64 | |
XM_006719471.3 | XP_006719534.1 | 4 - 5 | 449 | 64 | |
GenBank mRNA | BC022470.1 | - | 6 - 7 | 675 | 64 |
U48707.1 | - | 6 - 7 | 512 | 64 |