The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184965.1 | NP_001171894.1 | 11 - 12 | 2104 | 125 |
NM_001184966.1 | NP_001171895.1 | 10 - 11 | 2029 | 125 | |
NM_019045.4 | NP_061918.3 | 11 - 12 | 2104 | 125 | |
XM_011531353.2 | XP_011529655.1 | 10 - 11 | 2105 | 125 | |
XM_017029600.1 | XP_016885089.1 | 10 - 11 | 2037 | 125 | |
GenBank mRNA | AK001978.1 | - | 5 - 6 | 572 | 125 |
AK092077.1 | - | 8 - 9 | 1108 | 125 | |
AK127556.1 | - | 11 - 12 | 2015 | 125 | |
AK299711.1 | - | 10 - 11 | 1965 | 125 | |
AK304432.1 | - | 11 - 12 | 1631 | 125 | |
AL833605.1 | - | 11 - 12 | 2069 | 125 | |
AL834210.1 | - | 8 - 9 | 1371 | 125 | |
BC028697.2 | - | 11 - 12 | 2040 | 125 | |
BX538087.1 | - | 11 - 12 | 2046 | 125 | |
BX648150.1 | - | 11 - 12 | 2043 | 125 |