Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR44
Entrez Gene ID
54521
Gene Name
WD repeat domain 44
Gene Aliases
RAB11BP, RPH11
Chromosome Location
Chr.X: 118345997 - 118449961 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184965.1 NP_001171894.1 12 - 13 2166 80
NM_001184966.1 NP_001171895.1 11 - 12 2091 80
NM_019045.4 NP_061918.3 12 - 13 2166 80
XM_011531353.2 XP_011529655.1 11 - 12 2167 80
XM_017029600.1 XP_016885089.1 11 - 12 2099 80
GenBank mRNA AK001978.1 - 6 - 7 634 80
AK092077.1 - 9 - 10 1170 80
AK127556.1 - 12 - 13 2077 80
AK299711.1 - 11 - 12 2027 80
AK304432.1 - 12 - 13 1693 80
AL833605.1 - 12 - 13 2131 80
AL834210.1 - 9 - 10 1433 80
BC028697.2 - 12 - 13 2102 80
BX538087.1 - 12 - 13 2108 80
BX648150.1 - 12 - 13 2105 80

View Full Product Details