Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR44
Entrez Gene ID
54521
Gene Name
WD repeat domain 44
Gene Aliases
RAB11BP, RPH11
Chromosome Location
Chr.X: 118345997 - 118449961 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184965.1 NP_001171894.1 19 - 20 3050 94
NM_001184966.1 NP_001171895.1 17 - 18 2807 94
NM_019045.4 NP_061918.3 19 - 20 3074 94
XM_011531353.2 XP_011529655.1 18 - 19 3075 94
XM_017029600.1 XP_016885089.1 18 - 19 3007 94
GenBank mRNA AK001978.1 - 13 - 14 1535 94
AK092077.1 - 16 - 17 2078 94
AK299711.1 - 17 - 18 2743 94
AK304432.1 - 19 - 20 2601 94
AL833605.1 - 19 - 20 3015 94
BC028697.2 - 19 - 20 3010 94
BX537578.1 - 19 - 20 3048 94
BX538087.1 - 19 - 20 3016 94
BX648150.1 - 19 - 20 3013 94

View Full Product Details