The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184965.1 | NP_001171894.1 | 2 - 3 | 542 | 151 |
NM_019045.4 | NP_061918.3 | 2 - 3 | 542 | 151 | |
GenBank mRNA | AK127556.1 | - | 2 - 3 | 536 | 151 |
AK304432.1 | - | 2 - 3 | 69 | 151 | |
AL833605.1 | - | 2 - 3 | 507 | 151 | |
BC028697.2 | - | 2 - 3 | 478 | 151 | |
BX470418.1 | - | 2 - 3 | 543 | 151 | |
BX537578.1 | - | 2 - 3 | 517 | 151 | |
BX538087.1 | - | 2 - 3 | 484 | 151 | |
BX648150.1 | - | 2 - 3 | 481 | 151 |