The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289119.1 | NP_001276048.1 | 5 - 6 | 1132 | 75 |
NM_001289120.1 | NP_001276049.1 | 5 - 6 | 1155 | 75 | |
NM_001289121.1 | NP_001276050.1 | 5 - 6 | 1273 | 75 | |
NM_032226.2 | NP_115602.2 | 5 - 6 | 1128 | 75 | |
NR_110317.1 | 4 - 5 | 497 | 75 | ||
XM_005251608.4 | XP_005251665.1 | 5 - 6 | 2037 | 75 | |
XM_005251612.2 | XP_005251669.1 | 5 - 6 | 1095 | 75 | |
XM_011518051.2 | XP_011516353.1 | 4 - 5 | 6060 | 75 | |
XM_017015199.1 | XP_016870688.1 | 4 - 5 | 1053 | 75 | |
XM_017015200.1 | XP_016870689.1 | 4 - 5 | 1326 | 75 | |
XM_017015201.1 | XP_016870690.1 | 4 - 5 | 472 | 75 | |
GenBank mRNA | AK026264.1 | - | 5 - 6 | 1018 | 75 |
AK074608.1 | - | 4 - 5 | 75 | ||
AK303193.1 | - | 4 - 5 | 406 | 75 | |
AK315128.1 | - | 5 - 6 | 1145 | 75 | |
BC005990.1 | - | 5 - 6 | 1061 | 75 | |
BC036940.1 | - | 5 - 6 | 1246 | 75 |