The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001851.4 | NP_001842.3 | 28 - 29 | 2033 | 95 |
NM_078485.3 | NP_511040.2 | 22 - 23 | 1301 | 95 | |
XM_011535429.2 | XP_011533731.1 | 29 - 30 | 2063 | 95 | |
XM_011535430.2 | XP_011533732.1 | 23 - 24 | 1329 | 95 | |
XM_017010246.1 | XP_016865735.1 | 26 - 27 | 1420 | 95 | |
XM_017010247.1 | XP_016865736.1 | 20 - 21 | 1276 | 95 | |
GenBank mRNA | AK125738.1 | - | 18 - 19 | 966 | 95 |
BC008620.1 | - | 22 - 23 | 1228 | 95 | |
BC063646.1 | - | 22 - 23 | 1275 | 95 | |
X54412.1 | - | 28 - 29 | 2005 | 95 |